Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTTCTTTTTCTTAAACCTA[C/T]CCATTATATGTTAGTGTTCTTCTTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609512 | |||||||||||||||||||||||
Literature Links: |
CHMP2B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CHMP2B - charged multivesicular body protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244644.1 | Intron | NP_001231573.1 | ||||
NM_014043.3 | Intron | NP_054762.2 | ||||
XM_011533576.2 | Intron | XP_011531878.1 |
MIR4795 - microRNA 4795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |