Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTATATTAAATGTACATGTTTGC[A/G]GTAAATTACACCCTCATTTAAGAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609896 | |||||||||||||||||||||||
Literature Links: |
EIF4E3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EIF4E3 - eukaryotic translation initiation factor 4E family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134649.2 | 3532 | UTR 3 | NP_001128121.1 | |||
NM_001134650.1 | 3532 | UTR 3 | NP_001128122.1 | |||
NM_001134651.1 | 3532 | UTR 3 | NP_001128123.1 | |||
NM_001282886.1 | 3532 | UTR 3 | NP_001269815.1 | |||
NM_173359.4 | 3532 | UTR 3 | NP_775495.1 | |||
XM_011533651.2 | 3532 | UTR 3 | XP_011531953.1 | |||
XM_017006278.1 | 3532 | UTR 3 | XP_016861767.1 | |||
XM_017006279.1 | 3532 | UTR 3 | XP_016861768.1 | |||
XM_017006280.1 | 3532 | UTR 3 | XP_016861769.1 | |||
XM_017006281.1 | 3532 | UTR 3 | XP_016861770.1 | |||
XM_017006282.1 | 3532 | UTR 3 | XP_016861771.1 |