Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGCTTCCTGCCGTGCCTGGAAC[A/C]TTAGGGTGCTGGTGGCTGTCGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612307 | ||||||||||||||||||||
Literature Links: |
ADGRG7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRG7 - adhesion G protein-coupled receptor G7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308362.1 | 281 | Intron | NP_001295291.1 | |||
NM_032787.2 | 281 | Missense Mutation | ATT,CTT | I,L 9 | NP_116176.2 | |
XM_011513245.2 | 281 | UTR 5 | XP_011511547.1 |