Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACTAGGAGGATGTGCATGTTGA[A/G]TGTTTCCTTCTAGGTCCCATTGAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 190182 | |||||||||||||||||||||||
Literature Links: |
TGFBR2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TGFBR2 - transforming growth factor beta receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024847.2 | Intron | NP_001020018.1 | ||||
NM_003242.5 | Intron | NP_003233.4 | ||||
XM_011534043.2 | Intron | XP_011532345.1 | ||||
XM_011534045.2 | Intron | XP_011532347.1 | ||||
XM_017007106.1 | Intron | XP_016862595.1 |