Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCAGCACGAGGCCGAATCCCCC[A/G]TTGATCACCTCACCCCTGCAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613012 MIM: 615746 | ||||||||||||||||||||
Literature Links: |
UROC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UROC1 - urocanate hydratase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165974.1 | 1860 | Silent Mutation | AAC,AAT | N,N 662 | NP_001159446.1 | |
NM_144639.2 | 1860 | Silent Mutation | AAC,AAT | N,N 602 | NP_653240.1 |
ZXDC - ZXD family zinc finger C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |