Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTGGACCGCCCTACCAGGATC[C/T]GCTTCTCAGACTCCAATGGCAGTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603475 MIM: 600044 | |||||||||||||||||||||||
Literature Links: |
CHRD PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CHRD - chordin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304472.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | NP_001291401.1 | |
NM_001304473.1 | 880 | UTR 5 | NP_001291402.1 | |||
NM_001304474.1 | 880 | UTR 5 | NP_001291403.1 | |||
NM_003741.3 | 880 | Missense Mutation | CGC,TGC | R,C 212 | NP_003732.2 | |
XM_011513254.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_011511556.1 | |
XM_011513256.1 | 880 | UTR 5 | XP_011511558.1 | |||
XM_017007388.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_016862877.1 | |
XM_017007389.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_016862878.1 | |
XM_017007390.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_016862879.1 | |
XM_017007391.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_016862880.1 | |
XM_017007392.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_016862881.1 | |
XM_017007393.1 | 880 | Missense Mutation | CGC,TGC | R,C 212 | XP_016862882.1 | |
XM_017007394.1 | 880 | UTR 5 | XP_016862883.1 |
THPO - thrombopoietin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |