Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCGGAGGTAGTCGGCCATGCGC[C/T]GGGGCAGTGGCAGGCAGTCCACGTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602441 MIM: 602130 | |||||||||||||||||||||||
Literature Links: |
CISH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CISH - cytokine inducible SH2 containing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013324.5 | 3737 | Missense Mutation | CAG,CGG | Q,R 262 | NP_037456.5 | |
NM_145071.2 | 3737 | Missense Mutation | CAG,CGG | Q,R 245 | NP_659508.1 | |
XM_011533329.2 | 3737 | Missense Mutation | CAG,CGG | Q,R 251 | XP_011531631.1 |
MAPKAPK3 - mitogen-activated protein kinase-activated protein kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |