Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTTTATAGGTAGTTTTATAAAT[C/T]TGGGGATTTTCAAAAGCCACAAGAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 116947 | |||||||||||||||||||||||
Literature Links: |
CDC25A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CDC25A - cell division cycle 25A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001789.2 | Intron | NP_001780.2 | ||||
NM_201567.1 | Intron | NP_963861.1 | ||||
XM_006713434.2 | Intron | XP_006713497.1 | ||||
XM_006713435.2 | Intron | XP_006713498.1 | ||||
XM_011534316.1 | Intron | XP_011532618.1 |