Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTATCTTCAGGTGGACAGAGG[C/T]CCAGTGAGGACTGTGACTAAGCGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607726 | |||||||||||||||||||||||
Literature Links: |
PARP3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PARP3 - poly(ADP-ribose) polymerase family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003931.3 | 924 | Silent Mutation | GGC,GGT | G,G 178 | NP_001003931.3 | |
NM_005485.5 | 924 | Silent Mutation | GGC,GGT | G,G 171 | NP_005476.4 | |
XM_005264779.4 | 924 | Silent Mutation | GGC,GGT | G,G 171 | XP_005264836.2 | |
XM_017005490.1 | 924 | Silent Mutation | GGC,GGT | G,G 171 | XP_016860979.1 |
RRP9 - ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |