Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACTGTCGCACCCATGAGCAGGC[A/G]GTGGCGACACTGATCCACTAGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607910 | ||||||||||||||||||||
Literature Links: |
KIF9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF9 - kinesin family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134878.1 | 2422 | Missense Mutation | CGC,TGC | R,C 697 | NP_001128350.1 | |
NM_022342.4 | 2422 | Missense Mutation | CGC,TGC | R,C 632 | NP_071737.1 | |
NM_182902.3 | 2422 | Missense Mutation | CGC,TGC | R,C 697 | NP_878905.2 | |
XM_005265388.4 | 2422 | Missense Mutation | CGC,TGC | R,C 730 | XP_005265445.1 | |
XM_005265389.4 | 2422 | Missense Mutation | CGC,TGC | R,C 665 | XP_005265446.1 | |
XM_006713291.3 | 2422 | Missense Mutation | CGC,TGC | R,C 655 | XP_006713354.1 | |
XM_011534003.2 | 2422 | Missense Mutation | CGC,TGC | R,C 711 | XP_011532305.1 | |
XM_011534004.2 | 2422 | Missense Mutation | CGC,TGC | R,C 697 | XP_011532306.1 | |
XM_011534005.2 | 2422 | Missense Mutation | CGC,TGC | R,C 622 | XP_011532307.1 | |
XM_011534006.2 | 2422 | Missense Mutation | CGC,TGC | R,C 604 | XP_011532308.1 | |
XM_011534007.2 | 2422 | Missense Mutation | CGC,TGC | R,C 374 | XP_011532309.1 | |
XM_017007029.1 | 2422 | Missense Mutation | CGC,TGC | R,C 697 | XP_016862518.1 | |
XM_017007030.1 | 2422 | Missense Mutation | CGC,TGC | R,C 622 | XP_016862519.1 | |
XM_017007031.1 | 2422 | Missense Mutation | CGC,TGC | R,C 557 | XP_016862520.1 |
KIF9-AS1 - KIF9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |