Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAAAACATTATGGCTAAGAGAC[G/T]GGGAAGAAAGCTGCAGAAAAGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614802 MIM: 604944 | ||||||||||||||||||||
Literature Links: |
MSL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSL2 - male-specific lethal 2 homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145417.1 | 1659 | Missense Mutation | CAG,CCG | Q,P 235 | NP_001138889.1 | |
NM_018133.3 | 1659 | Missense Mutation | CAG,CCG | Q,P 309 | NP_060603.2 | |
XM_005247571.3 | 1659 | Missense Mutation | CAG,CCG | Q,P 235 | XP_005247628.1 | |
XM_006713684.3 | 1659 | Missense Mutation | CAG,CCG | Q,P 235 | XP_006713747.1 | |
XM_011512949.2 | 1659 | Missense Mutation | CAG,CCG | Q,P 235 | XP_011511251.1 |
PPP2R3A - protein phosphatase 2 regulatory subunit B''alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |