Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCCCAAGGAGGCGGTGGGCGC[C/G]GAGCAACCGCCCTCTCCTGCATCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602317 | |||||||||||||||||||||||
Literature Links: |
STAC PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
STAC - SH3 and cysteine rich domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292049.1 | 255 | Silent Mutation | GCC,GCG | A,A 22 | NP_001278978.1 | |
NM_003149.2 | 255 | Silent Mutation | GCC,GCG | A,A 22 | NP_003140.1 | |
XM_011534036.2 | 255 | UTR 5 | XP_011532338.1 | |||
XM_011534037.2 | 255 | Silent Mutation | GCC,GCG | A,A 22 | XP_011532339.1 | |
XM_011534038.2 | 255 | UTR 5 | XP_011532340.1 | |||
XM_017007083.1 | 255 | Intron | XP_016862572.1 | |||
XM_017007084.1 | 255 | Intron | XP_016862573.1 |