Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGGCACTAAGGGACTGATAAG[C/G]GGGTATTTTCTGTCTCTGGCAGTAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604419 MIM: 609381 | |||||||||||||||||||||||
Literature Links: |
POLQ PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
POLQ - DNA polymerase theta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199420.3 | 8638 | UTR 3 | NP_955452.3 | |||
XM_011512343.2 | 8638 | UTR 3 | XP_011510645.1 | |||
XM_011512347.2 | 8638 | UTR 3 | XP_011510649.1 | |||
XM_011512348.2 | 8638 | UTR 3 | XP_011510650.1 | |||
XM_011512352.2 | 8638 | Intron | XP_011510654.1 | |||
XM_011512354.2 | 8638 | UTR 3 | XP_011510656.1 | |||
XM_017005565.1 | 8638 | UTR 3 | XP_016861054.1 |
STXBP5L - syntaxin binding protein 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |