Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGTTACAGCATCTGCTGCTGA[A/G]AAACCAATCTGCAATATAAAAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603690 | ||||||||||||||||||||
Literature Links: |
SLC33A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC33A1 - solute carrier family 33 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190992.1 | 1517 | Silent Mutation | TTC,TTT | F,F 324 | NP_001177921.1 | |
NM_004733.3 | 1517 | Silent Mutation | TTC,TTT | F,F 324 | NP_004724.1 | |
XM_011513311.2 | 1517 | Intron | XP_011511613.1 | |||
XM_017007462.1 | 1517 | Missense Mutation | CTC,TTC | L,F 262 | XP_016862951.1 | |
XM_017007463.1 | 1517 | Silent Mutation | TTC,TTT | F,F 154 | XP_016862952.1 | |
XM_017007464.1 | 1517 | Silent Mutation | TTC,TTT | F,F 53 | XP_016862953.1 |