Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGTACATATATATTCAGAAATGT[C/T]TTGCCACCTGAGACCTATGAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TCAIM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCAIM - T-cell activation inhibitor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029839.2 | 432 | Missense Mutation | TCT,TTT | S,F 2 | NP_001025010.1 | |
NM_001029840.2 | 432 | Missense Mutation | TCT,TTT | S,F 2 | NP_001025011.1 | |
NM_001282913.1 | 432 | Missense Mutation | TCT,TTT | S,F 2 | NP_001269842.1 | |
NM_001282914.1 | 432 | UTR 5 | NP_001269843.1 | |||
NM_001282915.1 | 432 | UTR 5 | NP_001269844.1 | |||
NM_173826.3 | 432 | Missense Mutation | TCT,TTT | S,F 2 | NP_776187.2 | |
XM_017006251.1 | 432 | UTR 5 | XP_016861740.1 |