Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTAACAATGACTGTCTATGATGT[C/G]TCAGTTCTTATCTGAATTCCAAAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609262 MIM: 612907 | |||||||||||||||||||||||
Literature Links: |
CRBN PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CRBN - cereblon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173482.1 | 3515 | Intron | NP_001166953.1 | |||
NM_016302.3 | 3515 | Intron | NP_057386.2 | |||
XM_005265202.3 | 3515 | Intron | XP_005265259.1 | |||
XM_011533791.2 | 3515 | UTR 3 | XP_011532093.1 | |||
XM_011533793.2 | 3515 | Intron | XP_011532095.1 | |||
XM_011533794.2 | 3515 | Intron | XP_011532096.1 |
TRNT1 - tRNA nucleotidyl transferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302946.1 | 3515 | UTR 3 | NP_001289875.1 | |||
NM_182916.2 | 3515 | UTR 3 | NP_886552.2 | |||
XM_011533776.2 | 3515 | UTR 3 | XP_011532078.1 | |||
XM_011533777.2 | 3515 | UTR 3 | XP_011532079.1 | |||
XM_011533778.2 | 3515 | Intron | XP_011532080.1 |