Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATGATCGGAGGGGTGATAGATA[C/T]GATGACTACCGAGACTATGACAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606884 | ||||||||||||||||||||
Literature Links: |
RBM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM5 - RNA binding motif protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005778.3 | 328 | Silent Mutation | TAC,TAT | Y,Y 51 | NP_005769.1 | |
XM_006712917.1 | 328 | Silent Mutation | TAC,TAT | Y,Y 51 | XP_006712980.1 | |
XM_011533261.1 | 328 | Silent Mutation | TAC,TAT | Y,Y 51 | XP_011531563.1 | |
XM_011533262.1 | 328 | Silent Mutation | TAC,TAT | Y,Y 51 | XP_011531564.1 | |
XM_017005503.1 | 328 | Silent Mutation | TAC,TAT | Y,Y 51 | XP_016860992.1 | |
XM_017005504.1 | 328 | Silent Mutation | TAC,TAT | Y,Y 51 | XP_016860993.1 | |
XM_017005505.1 | 328 | UTR 5 | XP_016860994.1 | |||
XM_017005506.1 | 328 | UTR 5 | XP_016860995.1 | |||
XM_017005507.1 | 328 | UTR 5 | XP_016860996.1 |
RBM5-AS1 - RBM5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |