Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAAGGAAATCTATGAGAACATG[A/G]CCCCTGGGGAGAATAAGGTAAAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610145 MIM: 606223 | ||||||||||||||||||||
Literature Links: |
ECE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ECE2 - endothelin converting enzyme 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD2 - proteasome 26S subunit, non-ATPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278708.1 | 509 | Intron | NP_001265637.1 | |||
NM_001278709.1 | 509 | Intron | NP_001265638.1 | |||
NM_002808.4 | 509 | Missense Mutation | ACC,GCC | T,A 114 | NP_002799.3 |