Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTCCATTGCCAATCCCTTTTGG[A/T]CAGGACCATGGTCCTCTGAAAGAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608753 | |||||||||||||||||||||||
Literature Links: |
TSEN2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TSEN2 - tRNA splicing endonuclease subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145392.1 | 401 | Intron | NP_001138864.1 | |||
NM_001145393.2 | 401 | Intron | NP_001138865.1 | |||
NM_001145394.1 | 401 | Intron | NP_001138866.1 | |||
NM_001321277.1 | 401 | Intron | NP_001308206.1 | |||
NM_001321278.1 | 401 | Silent Mutation | GGA,GGT | G,G 28 | NP_001308207.1 | |
NM_001321279.1 | 401 | Silent Mutation | GGA,GGT | G,G 28 | NP_001308208.1 | |
NM_025265.3 | 401 | Silent Mutation | GGA,GGT | G,G 28 | NP_079541.1 | |
XM_011534139.2 | 401 | Intron | XP_011532441.1 | |||
XM_017007292.1 | 401 | Intron | XP_016862781.1 | |||
XM_017007293.1 | 401 | Silent Mutation | GGA,GGT | G,G 28 | XP_016862782.1 | |
XM_017007294.1 | 401 | Intron | XP_016862783.1 | |||
XM_017007295.1 | 401 | Silent Mutation | GGA,GGT | G,G 28 | XP_016862784.1 | |
XM_017007296.1 | 401 | Silent Mutation | GGA,GGT | G,G 28 | XP_016862785.1 | |
XM_017007297.1 | 401 | Silent Mutation | GGA,GGT | G,G 28 | XP_016862786.1 |