Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATGAAATCAAGAAGCTGATCTA[C/T]GTACATCTGGTCATATGGCTGCTGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614995 MIM: 616012 | |||||||||||||||||||||||
Literature Links: |
IL17RE PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IL17RE - interleukin 17 receptor E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JAGN1 - jagunal homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032492.3 | 262 | Silent Mutation | TAC,TAT | Y,Y 42 | NP_115881.3 |