Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGCAGTATCTATTCACAATGAA[A/G]TTGCAGTCTCCCGAATTCCAGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612907 | ||||||||||||||||||||
Literature Links: |
TRNT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRNT1 - tRNA nucleotidyl transferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302946.1 | 441 | Silent Mutation | AAA,AAG | K,K 31 | NP_001289875.1 | |
NM_182916.2 | 441 | Silent Mutation | AAA,AAG | K,K 31 | NP_886552.2 | |
XM_011533776.2 | 441 | Silent Mutation | AAA,AAG | K,K 31 | XP_011532078.1 | |
XM_011533777.2 | 441 | Silent Mutation | AAA,AAG | K,K 31 | XP_011532079.1 | |
XM_011533778.2 | 441 | Silent Mutation | AAA,AAG | K,K 31 | XP_011532080.1 |