Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGATGCAGATGCGCTTCGATG[A/G]ACGCCTGGGCTTCCCCGGCGGATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC339874 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC339874 - uncharacterized LOC339874 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT16 - nudix hydrolase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171905.1 | 95 | Missense Mutation | GAA,GGA | E,G 7 | NP_001165376.1 | |
NM_001171906.1 | 95 | Missense Mutation | GAA,GGA | E,G 53 | NP_001165377.1 | |
NM_152395.2 | 95 | Missense Mutation | GAA,GGA | E,G 53 | NP_689608.2 |