Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCTCGCTGGGTGGACCAGCAGC[A/G]TTTTCTGCTCGGCATTCCCAAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165340 | ||||||||||||||||||||
Literature Links: |
SKIL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SKIL - SKI-like proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145097.2 | 1030 | Silent Mutation | GCA,GCG | A,A 106 | NP_001138569.1 | |
NM_001145098.2 | 1030 | Silent Mutation | GCA,GCG | A,A 86 | NP_001138570.1 | |
NM_001248008.1 | 1030 | Silent Mutation | GCA,GCG | A,A 106 | NP_001234937.1 | |
NM_005414.4 | 1030 | Silent Mutation | GCA,GCG | A,A 106 | NP_005405.2 | |
XM_005247721.1 | 1030 | Silent Mutation | GCA,GCG | A,A 106 | XP_005247778.1 | |
XM_006713735.1 | 1030 | Silent Mutation | GCA,GCG | A,A 106 | XP_006713798.1 |