Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGCTGGTCAGTGTGGCAATTCTC[C/T]GCAGGTTTTCCCTGACTTTAATAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608295 | ||||||||||||||||||||
Literature Links: |
FAM107A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM107A - family with sequence similarity 107 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076778.2 | 1104 | Missense Mutation | CAG,CGG | Q,R 132 | NP_001070246.1 | |
NM_001282713.1 | 1104 | Missense Mutation | CAG,CGG | Q,R 160 | NP_001269642.1 | |
NM_001282714.1 | 1104 | Missense Mutation | CAG,CGG | Q,R 163 | NP_001269643.1 | |
NM_007177.3 | 1104 | Missense Mutation | CAG,CGG | Q,R 132 | NP_009108.1 |