Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGTTAGAGAGTGCCATGGAGC[A/G]GTGCAGCACATAGCTCCGATAGGCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604427 | |||||||||||||||||||||||
Literature Links: |
SCN10A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SCN10A - sodium voltage-gated channel alpha subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293306.2 | 5311 | Missense Mutation | CGC,TGC | R,C 1868 | NP_001280235.2 | |
NM_001293307.2 | 5311 | Missense Mutation | CGC,TGC | R,C 1771 | NP_001280236.2 | |
NM_006514.3 | 5311 | Missense Mutation | CGC,TGC | R,C 1869 | NP_006505.3 | |
XM_005265371.3 | 5311 | Missense Mutation | CGC,TGC | R,C 1872 | XP_005265428.1 | |
XM_011533993.2 | 5311 | Missense Mutation | CGC,TGC | R,C 1871 | XP_011532295.1 | |
XM_011533994.2 | 5311 | Missense Mutation | CGC,TGC | R,C 1774 | XP_011532296.1 |