Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACAGGAAAGCCAAATCCAACA[C/T]TGATTCTGCCAAAATCTTTAGTAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF445 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF445 - zinc finger protein 445 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181489.5 | 5884 | UTR 3 | NP_852466.1 | |||
XM_005265102.2 | 5884 | UTR 3 | XP_005265159.1 | |||
XM_005265103.3 | 5884 | UTR 3 | XP_005265160.1 | |||
XM_011533674.2 | 5884 | UTR 3 | XP_011531976.1 | |||
XM_011533675.2 | 5884 | UTR 3 | XP_011531977.1 | |||
XM_017006345.1 | 5884 | UTR 3 | XP_016861834.1 | |||
XM_017006346.1 | 5884 | UTR 3 | XP_016861835.1 |