Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCTTGACCTTGCTGGCCTTCT[C/G]AAAGCAGTCAGCATCAGGTCCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607474 | ||||||||||||||||||||
Literature Links: |
HGD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HGD - homogentisate 1,2-dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000187.3 | 1595 | Missense Mutation | CAG,GAG | Q,E 379 | NP_000178.2 | |
XM_005247412.1 | 1595 | Missense Mutation | CAG,GAG | Q,E 304 | XP_005247469.1 | |
XM_005247413.1 | 1595 | Intron | XP_005247470.1 | |||
XM_005247414.4 | 1595 | Intron | XP_005247471.1 | |||
XM_011512746.1 | 1595 | Intron | XP_011511048.1 | |||
XM_017006277.1 | 1595 | Missense Mutation | CAG,GAG | Q,E 238 | XP_016861766.1 |