Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTGGCCTCTGGGTCAAACTTCT[C/T]CCAGGTCTCATAGAACATGTCAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604427 | ||||||||||||||||||||
Literature Links: |
SCN10A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCN10A - sodium voltage-gated channel alpha subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293306.2 | 4951 | Missense Mutation | AAG,GAG | K,E 1748 | NP_001280235.2 | |
NM_001293307.2 | 4951 | Missense Mutation | AAG,GAG | K,E 1651 | NP_001280236.2 | |
NM_006514.3 | 4951 | Missense Mutation | AAG,GAG | K,E 1749 | NP_006505.3 | |
XM_005265371.3 | 4951 | Missense Mutation | AAG,GAG | K,E 1752 | XP_005265428.1 | |
XM_011533993.2 | 4951 | Missense Mutation | AAG,GAG | K,E 1751 | XP_011532295.1 | |
XM_011533994.2 | 4951 | Missense Mutation | AAG,GAG | K,E 1654 | XP_011532296.1 |