Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCTTGGAGGACGCTGTGGATAC[A/G]GTACACTGCTCAGAAGCAAAGAAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TMEM40 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMEM40 - transmembrane protein 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284406.1 | 840 | Missense Mutation | CGT,TGT | R,C 225 | NP_001271335.1 | |
NM_001284407.1 | 840 | Missense Mutation | CGT,TGT | R,C 179 | NP_001271336.1 | |
NM_001284408.1 | 840 | Missense Mutation | CGT,TGT | R,C 133 | NP_001271337.1 | |
NM_018306.3 | 840 | Missense Mutation | CGT,TGT | R,C 209 | NP_060776.2 | |
XM_011533937.2 | 840 | Missense Mutation | CGT,TGT | R,C 209 | XP_011532239.1 |