Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAGTCAGTCACCTTAACAAGAT[A/G]GGCAATTCTTGCTGGAGACTGAAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612767 | |||||||||||||||||||||||
Literature Links: |
DHX36 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DHX36 - DEAH-box helicase 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114397.1 | 2911 | Missense Mutation | CAT,TAT | H,Y 930 | NP_001107869.1 | |
NM_020865.2 | 2911 | Missense Mutation | CAT,TAT | H,Y 944 | NP_065916.2 |