Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCATATGACCTTGTGGCATTGA[A/G]TAGATATTCATAAAGTTGCTTAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604384 | ||||||||||||||||||||
Literature Links: |
ASTE1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASTE1 - asteroid homolog 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288950.1 | 2166 | Missense Mutation | CTC,TTC | L,F 633 | NP_001275879.1 | |
NM_014065.3 | 2166 | Missense Mutation | CTC,TTC | L,F 608 | NP_054784.2 | |
XM_017006266.1 | 2166 | UTR 3 | XP_016861755.1 | |||
XM_017006267.1 | 2166 | UTR 3 | XP_016861756.1 | |||
XM_017006268.1 | 2166 | UTR 3 | XP_016861757.1 | |||
XM_017006269.1 | 2166 | UTR 3 | XP_016861758.1 |
ATP2C1 - ATPase secretory pathway Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001485.2 | 2166 | Intron | NP_001001485.1 | |||
NM_001001486.1 | 2166 | Intron | NP_001001486.1 | |||
NM_001001487.1 | 2166 | Intron | NP_001001487.1 | |||
NM_001199179.1 | 2166 | Intron | NP_001186108.1 | |||
NM_001199180.1 | 2166 | Intron | NP_001186109.1 | |||
NM_001199181.1 | 2166 | Intron | NP_001186110.1 | |||
NM_001199182.1 | 2166 | Intron | NP_001186111.1 | |||
NM_001199183.1 | 2166 | Intron | NP_001186112.1 | |||
NM_001199184.1 | 2166 | Intron | NP_001186113.1 | |||
NM_001199185.1 | 2166 | Intron | NP_001186114.1 | |||
NM_014382.3 | 2166 | Intron | NP_055197.2 | |||
XM_005247354.2 | 2166 | Intron | XP_005247411.1 | |||
XM_005247355.2 | 2166 | Intron | XP_005247412.1 | |||
XM_005247356.2 | 2166 | Intron | XP_005247413.1 | |||
XM_005247358.2 | 2166 | Intron | XP_005247415.1 | |||
XM_011512686.2 | 2166 | Intron | XP_011510988.1 | |||
XM_017006164.1 | 2166 | Intron | XP_016861653.1 |