Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTGTCCACCTATCCTAGAACTT[A/G]GGAATGAAATAGGCTAGACTCAACT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 180220 | |||||||||||||||||||||||
Literature Links: |
RARB PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RARB - retinoic acid receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000965.4 | Intron | NP_000956.2 | ||||
NM_001290216.2 | Intron | NP_001277145.1 | ||||
NM_001290217.1 | Intron | NP_001277146.1 | ||||
NM_001290266.1 | Intron | NP_001277195.1 | ||||
NM_001290276.1 | Intron | NP_001277205.1 | ||||
NM_001290277.1 | Intron | NP_001277206.1 | ||||
NM_001290300.1 | Intron | NP_001277229.1 | ||||
NM_016152.3 | Intron | NP_057236.1 |