Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGCTGGATTGGCTGCTTGATGA[C/T]TGCTCTCTTTCTTTTGTAATTTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611177 | ||||||||||||||||||||
Literature Links: |
IFT80 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT80 - intraflagellar transport 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190241.1 | 2722 | Silent Mutation | CAA,CAG | Q,Q 626 | NP_001177170.1 | |
NM_001190242.1 | 2722 | Silent Mutation | CAA,CAG | Q,Q 626 | NP_001177171.1 | |
NM_020800.2 | 2722 | Silent Mutation | CAA,CAG | Q,Q 763 | NP_065851.1 |