Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCCTGGGGAGATTCGGACCTG[A/G]GTACAGCTCCATCACCCTGGAAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF197 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF197 - zinc finger protein 197 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264783.4 | 492 | Nonsense Mutation | TGA,TGG | *,W 101 | XP_005264840.1 | |
XM_017005495.1 | 492 | Nonsense Mutation | TGA,TGG | *,W 101 | XP_016860984.1 |
ZNF197-AS1 - ZNF197 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |