Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTTCTACCAGCGCTTTCCGCT[C/G]AGCTTCGGCTTCCAGCGTCTGAGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616596 MIM: 607673 | |||||||||||||||||||||||
Literature Links: |
ARL8B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARL8B - ADP ribosylation factor like GTPase 8B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EDEM1 - ER degradation enhancing alpha-mannosidase like protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014674.2 | 179 | Silent Mutation | CTC,CTG | L,L 39 | NP_055489.1 | |
XM_011534271.2 | 179 | Intron | XP_011532573.1 | |||
XM_011534272.2 | 179 | UTR 5 | XP_011532574.1 |