Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCACGGGGTAGATGCGGCTTG[C/T]TCCAGGGCCCAGCCAGTAGCCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610068 | ||||||||||||||||||||
Literature Links: |
SLC26A6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC26A6 - solute carrier family 26 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM89 - transmembrane protein 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008269.1 | 187 | Missense Mutation | ACA,GCA | T,A 63 | NP_001008270.1 |