Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGAGAAAAGAGCATCCAGAGA[G/T]GTTAGGGTTGACAGGATGGACCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604915 | ||||||||||||||||||||
Literature Links: |
MAP3K13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K13 - mitogen-activated protein kinase kinase kinase 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM41A - transmembrane protein 41A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080652.3 | 567 | Silent Mutation | ACA,ACC | T,T 211 | NP_542383.1 | |
XM_017007437.1 | 567 | Silent Mutation | ACA,ACC | T,T 157 | XP_016862926.1 |