Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCAAGAAGCGGGACGTTCACCA[C/T]GGGAAGAAAATCGCTGTACCTTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600338 | ||||||||||||||||||||
Literature Links: |
AADAC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AADAC - arylacetamide deacetylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001086.2 | 92 | Missense Mutation | ACG,ATG | T,M 1 | NP_001077.2 | |
XM_005247104.4 | 92 | Missense Mutation | ACG,ATG | T,M 1 | XP_005247161.1 |
AADACL2-AS1 - AADACL2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |