Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACTCCAACCATCACCACCACCA[A/C]AATGTTTCTGCGAGCACCACTCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602410 | ||||||||||||||||||||
Literature Links: |
BRPF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRPF1 - bromodomain and PHD finger containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003694.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | NP_001003694.1 | |
NM_001319049.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | NP_001305978.1 | |
NM_001319050.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | NP_001305979.1 | |
NM_004634.2 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | NP_004625.2 | |
XM_005265449.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | XP_005265506.1 | |
XM_005265450.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | XP_005265507.1 | |
XM_005265452.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | XP_005265509.1 | |
XM_011534101.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | XP_011532403.1 | |
XM_011534102.1 | 918 | Missense Mutation | CAA,CAC | Q,H 166 | XP_011532404.1 |
CPNE9 - copine family member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |