Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCCTGTGAAGGAGAGCTTCAAG[C/T]TGGCAAAGGAGATGAAGTCACAATT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608315 | |||||||||||||||||||||||
Literature Links: |
EAF1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EAF1 - ELL associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033083.6 | 378 | Missense Mutation | GCT,GTT | A,V 52 | NP_149074.3 | |
XM_005265518.1 | 378 | UTR 5 | XP_005265575.1 | |||
XM_011534165.1 | 378 | UTR 5 | XP_011532467.1 | |||
XM_011534166.1 | 378 | UTR 5 | XP_011532468.1 |
METTL6 - methyltransferase like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |