Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTCTGCCATGGCCACATTGGT[A/G]TTGAACTCATTCTGGTATTTCTATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604282 | ||||||||||||||||||||
Literature Links: |
H1FOO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
H1FOO - H1 histone family member O, oocyte specific | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLXND1 - plexin D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015103.2 | 5232 | Silent Mutation | AAC,AAT | N,N 1869 | NP_055918.2 | |
XM_011512588.1 | 5232 | Silent Mutation | AAC,AAT | N,N 1869 | XP_011510890.1 | |
XM_011512589.1 | 5232 | Silent Mutation | AAC,AAT | N,N 1739 | XP_011510891.1 | |
XM_011512590.2 | 5232 | Intron | XP_011510892.1 | |||
XM_011512592.1 | 5232 | Silent Mutation | AAC,AAT | N,N 925 | XP_011510894.1 |