Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGCCATTCTTCCAGGCGCGAGG[C/T]AGCAGCGGCTGCAGTTCAACATGAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607953 | |||||||||||||||||||||||
Literature Links: |
ABHD10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ABHD10 - abhydrolase domain containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAGLN3 - transgelin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008272.1 | 215 | UTR 5 | NP_001008273.1 | |||
NM_001008273.1 | 215 | Intron | NP_001008274.1 | |||
NM_013259.2 | 215 | UTR 5 | NP_037391.2 |