Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGTTCAACTTACCTTTGTCTG[C/T]TCTTTTCTAAGTTGTTTTAATAACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC58 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC58 - coiled-coil domain containing 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017928.3 | 372 | Intron | NP_001017928.1 | |||
NM_001308326.1 | 372 | Intron | NP_001295255.1 | |||
XM_011512410.2 | 372 | Intron | XP_011510712.1 | |||
XM_017005697.1 | 372 | Silent Mutation | GAA,GAG | E,E 105 | XP_016861186.1 |