Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATTCACATCTTTTTATACATC[G/T]TAAACAAGGATAGGATACTTCTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610412 | ||||||||||||||||||||
Literature Links: |
SPTSSB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPTSSB - serine palmitoyltransferase small subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040100.1 | 1442 | UTR 3 | NP_001035189.1 | |||
NM_001320679.1 | 1442 | UTR 3 | NP_001307608.1 | |||
XM_017005793.1 | 1442 | UTR 3 | XP_016861282.1 | |||
XM_017005794.1 | 1442 | UTR 3 | XP_016861283.1 |