Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACAGCTTGTATGTGATAGCATG[C/T]ATGCAAACCTTAATGCCTCCCAATG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF502 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF502 - zinc finger protein 502 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134440.1 | Intron | NP_001127912.1 | ||||
NM_001134441.1 | Intron | NP_001127913.1 | ||||
NM_001134442.1 | Intron | NP_001127914.1 | ||||
NM_001282880.1 | Intron | NP_001269809.1 | ||||
NM_033210.4 | Intron | NP_149987.2 | ||||
XM_017007440.1 | Intron | XP_016862929.1 |