Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCGTCAGACAGACACTGCTCC[C/G]GGCACGATCCTGCGGATGTGGGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614214 | ||||||||||||||||||||
Literature Links: |
KLHL6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL6 - kelch like family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130446.2 | 1561 | Silent Mutation | CCC,CCG | P,P 616 | NP_569713.2 | |
XM_011513273.2 | 1561 | Silent Mutation | CCC,CCG | P,P 489 | XP_011511575.1 | |
XM_011513274.2 | 1561 | Intron | XP_011511576.1 |