Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAACAATGACACGCTTGCCATCA[A/G]TAGGTACAATGGGCTTCTTCAATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116947 | ||||||||||||||||||||
Literature Links: |
CDC25A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC25A - cell division cycle 25A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001789.2 | 1715 | Missense Mutation | ACT,ATT | T,I 420 | NP_001780.2 | |
NM_201567.1 | 1715 | Missense Mutation | ACT,ATT | T,I 380 | NP_963861.1 | |
XM_006713434.2 | 1715 | Missense Mutation | ACT,ATT | T,I 419 | XP_006713497.1 | |
XM_006713435.2 | 1715 | Missense Mutation | ACT,ATT | T,I 379 | XP_006713498.1 | |
XM_011534316.1 | 1715 | Missense Mutation | ACT,ATT | T,I 420 | XP_011532618.1 |