Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGTGATCATAAAGTCTGACCA[A/G]TTCACAATGTTTCTTTTTCTAAGAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
C3orf67 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C3orf67 - chromosome 3 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198463.2 | Intron | NP_940865.1 | ||||
XM_005264929.2 | Intron | XP_005264986.1 | ||||
XM_006713013.3 | Intron | XP_006713076.1 | ||||
XM_011533451.2 | Intron | XP_011531753.1 | ||||
XM_011533453.2 | Intron | XP_011531755.1 | ||||
XM_011533455.2 | Intron | XP_011531757.1 | ||||
XM_011533456.1 | Intron | XP_011531758.1 | ||||
XM_011533457.1 | Intron | XP_011531759.1 | ||||
XM_011533458.2 | Intron | XP_011531760.1 | ||||
XM_011533459.2 | Intron | XP_011531761.1 | ||||
XM_011533460.2 | Intron | XP_011531762.1 | ||||
XM_011533461.1 | Intron | XP_011531763.1 | ||||
XM_011533463.2 | Intron | XP_011531765.1 | ||||
XM_017005848.1 | Intron | XP_016861337.1 | ||||
XM_017005849.1 | Intron | XP_016861338.1 | ||||
XM_017005850.1 | Intron | XP_016861339.1 | ||||
XM_017005851.1 | Intron | XP_016861340.1 |