Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCCCCCCTTCCGCACCTTCTC[A/G]GCCAGCGACTGGGGCCTCACCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601055 | ||||||||||||||||||||
Literature Links: |
PLXNA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLXNA1 - plexin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032242.3 | 830 | Silent Mutation | TCA,TCG | S,S 45 | NP_115618.3 | |
XM_011512908.2 | 830 | Silent Mutation | TCA,TCG | S,S 63 | XP_011511210.1 | |
XM_011512909.1 | 830 | Silent Mutation | TCA,TCG | S,S 63 | XP_011511211.1 |