Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTTGGGCCTGGTATGTACAGA[G/T]GGCTGGTTAGGATTCTCGGCACCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608261 | ||||||||||||||||||||
Literature Links: |
SENP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SENP2 - SUMO1/sentrin/SMT3 specific peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021627.2 | 127 | Missense Mutation | GGG,TGG | G,W 4 | NP_067640.2 | |
XM_005247690.2 | 127 | Missense Mutation | GGG,TGG | G,W 4 | XP_005247747.2 | |
XM_005247691.2 | 127 | UTR 5 | XP_005247748.1 | |||
XM_017006973.1 | 127 | UTR 5 | XP_016862462.1 |